ENAH38A2DAFD Enamine

(6-chloro-4-methylpyridin-3-yl)methanamine

  • MDL number MFCD18254319
  • Cas Number : 1256824-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354377
Availability details for
Quantity
  • Availability
  • Add to Cart