ENAH38A2DB08 Enamine

(3aR,6aS)-2-oxo-hexahydrocyclopenta[d][1,3]dioxole-5-carboxylic acid

  • MDL number MFCD31915367
  • Cas Number : 2309431-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354429
Availability details for
Quantity
  • Availability
  • Add to Cart