ENAH38A2DB0E Enamine

(2E)-3-(3-fluoro-4-methoxyphenyl)-2-methylprop-2-enoic acid

  • MDL number MFCD06207001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354463
Availability details for
Quantity
  • Availability
  • Add to Cart