ENAH38A2DB12 Enamine

6-(4-bromophenyl)-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD05881523
  • Cas Number : 791786-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354511
Availability details for
Quantity
  • Availability
  • Add to Cart