ENAH38A2DB19 Enamine

2,3,4,5-tetrahydro-1-benzothiepin-4-one

  • MDL number MFCD24689953
  • Cas Number : 65039-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354527
Availability details for
Quantity
  • Availability
  • Add to Cart