ENAH38A2DB29 Enamine

6-(aminomethyl)-6-methylpiperidin-2-one; trifluoroacetic acid

  • MDL number MFCD31915862
  • Cas Number : 2309456-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354697
Availability details for
Quantity
  • Availability
  • Add to Cart