ENAH38A2DB2E Enamine

(3E)-1-bromo-4-ethoxy-1,1-difluorobut-3-en-2-one

  • MDL number MFCD30345348

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354723
Availability details for
Quantity
  • Availability
  • Add to Cart