ENAH38A2DB31 Enamine

methyl 2-chloro-5-(chlorosulfonyl)thiophene-3-carboxylate

  • MDL number MFCD31915863
  • Cas Number : 1629319-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354749
Availability details for
Quantity
  • Availability
  • Add to Cart