ENAH38A2DB34 Enamine

methyl 2-[3-(aminomethyl)-1H-1,2,4-triazol-1-yl]acetate dihydrochloride

  • MDL number MFCD32066255
  • Cas Number : 2309447-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354778
Availability details for
Quantity
  • Availability
  • Add to Cart