ENAH38A2DB36 Enamine

1-(2-aminoethyl)piperidine-4-carboxylic acid dihydrochloride

  • MDL number MFCD27959742
  • Cas Number : 1417638-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354803
Availability details for
Quantity
  • Availability
  • Add to Cart