ENAH38A2DB3B Enamine

2-amino-2-[3-(methoxycarbonyl)phenyl]acetic acid; trifluoroacetic acid

  • MDL number MFCD31915432
  • Cas Number : 2309444-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354818
Availability details for
Quantity
  • Availability
  • Add to Cart