ENAH38A2DB42 Enamine

(2E)-4-{[(tert-butoxy)carbonyl]amino}pent-2-enoic acid

  • MDL number MFCD20232511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354830
Availability details for
Quantity
  • Availability
  • Add to Cart