ENAH38A2DB4C Enamine

methyl 2-(6-amino-5,6,7,8-tetrahydronaphthalen-2-yl)acetate hydrochloride

  • MDL number MFCD31915153
  • Cas Number : 2309457-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354860
Availability details for
Quantity
  • Availability
  • Add to Cart