ENAH38A2DB52 Enamine

(1r,3r)-3-(phenoxymethyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD31915157
  • Cas Number : 2309446-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354990
Availability details for
Quantity
  • Availability
  • Add to Cart