ENAH38A2DB53 Enamine

3-[(3-chloro-4-fluorophenyl)amino]pyridazine-4-carboxylic acid

  • MDL number MFCD24984567
  • Cas Number : 1539785-66-4
  • Molecular Weight : 267.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7354994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354994
Availability details for
Quantity
  • Availability
  • Add to Cart