ENAH38A2DB56 Enamine

2-{4-[(tert-butoxy)carbonyl]-5,5-dimethylmorpholin-2-yl}acetic acid

  • MDL number MFCD06657467
  • Cas Number : 889953-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354997
Availability details for
Quantity
  • Availability
  • Add to Cart