ENAH38A2DB5C Enamine

methyl decahydroisoquinoline-6-carboxylate

  • MDL number MFCD24444641
  • Cas Number : 104654-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355011
Availability details for
Quantity
  • Availability
  • Add to Cart