ENAH38A2DB67 Enamine

tert-butyl (2S,4S)-2-(aminomethyl)-4-(trifluoromethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD31742658
  • Cas Number : 2165957-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355059
Availability details for
Quantity
  • Availability
  • Add to Cart