ENAH38A2DB76 Enamine

(1R)-1-(3,3-difluorocyclobutyl)ethan-1-amine hydrochloride

  • MDL number MFCD31915223
  • Cas Number : 1621223-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355125
Availability details for
Quantity
  • Availability
  • Add to Cart