ENAH38A2DB7A Enamine

3-bromo-N,N-dimethyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD22395552
  • Cas Number : 1824339-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355155
Availability details for
Quantity
  • Availability
  • Add to Cart