ENAH38A2DB81 Enamine

3-(2,2-difluoroethoxy)-1H-pyrazole hydrochloride

  • MDL number MFCD31915227
  • Cas Number : 2309475-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355178
Availability details for
Quantity
  • Availability
  • Add to Cart