ENAH5802D9BB Enamine

1-(but-3-yn-1-yl)azocane hydrochloride

  • MDL number MFCD32069015
  • Cas Number : 2375269-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437216
Availability details for
Quantity
  • Availability
  • Add to Cart