ENAH5802D9BE Enamine

1-(but-3-yn-1-yl)-3,3-diethylpyrrolidine hydrochloride

  • MDL number MFCD32068990
  • Cas Number : 2375259-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437473
Availability details for
Quantity
  • Availability
  • Add to Cart