ENAH5802D9BF Enamine

[1-(but-3-yn-1-yl)-3-methoxypiperidin-3-yl]methanol hydrochloride

  • MDL number MFCD32069072
  • Cas Number : 2375258-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437792
Availability details for
Quantity
  • Availability
  • Add to Cart