ENAH5802D9C0 Enamine

(1,2,3,6-tetrahydropyridin-2-yl)methanol hydrochloride

  • Cas Number : 2375262-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439328
Availability details for
Quantity
  • Availability
  • Add to Cart