ENAH5802D9C3 Enamine

(1-cyclopropyl-3-methylazetidin-3-yl)methanol

  • MDL number MFCD32068883
  • Cas Number : 2375259-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717138
Availability details for
Quantity
  • Availability
  • Add to Cart