ENAH5802D9C6 Enamine

(1s,3s)-3-amino-1-cyclopentylcyclobutan-1-ol hydrochloride

  • Cas Number : 2375258-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427339
Availability details for
Quantity
  • Availability
  • Add to Cart