ENAH5802D9D1 Enamine

(1R)-1-(2,5-difluorophenyl)-2,2-difluoroethan-1-ol

  • MDL number MFCD32069393
  • Cas Number : 2375250-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435385
Availability details for
Quantity
  • Availability
  • Add to Cart