ENAH5802D9D2 Enamine

(1S)-1-(2,5-difluorophenyl)-2,2-difluoroethan-1-ol

  • MDL number MFCD32069392
  • Cas Number : 2375249-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435384
Availability details for
Quantity
  • Availability
  • Add to Cart