ENAH5802D9D4 Enamine

(1R)-1-(2,3-difluorophenyl)-2,2-difluoroethan-1-ol

  • MDL number MFCD32068759
  • Cas Number : 2375249-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435224
Availability details for
Quantity
  • Availability
  • Add to Cart