ENAH5802D9D6 Enamine

[3-(1,1,1-trifluoro-2-methylpropan-2-yl)phenyl]methanol

  • MDL number MFCD32069187
  • Cas Number : 2375259-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438758
Availability details for
Quantity
  • Availability
  • Add to Cart