ENAH5802D9D7 Enamine

14-amino-3,6,9,12-tetraoxatetradecan-1-ol hydrochloride

  • MDL number MFCD32068834
  • Cas Number : 2375259-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427656
Availability details for
Quantity
  • Availability
  • Add to Cart