ENAH5802D9DB Enamine

tert-butyl 4-(hydroxymethyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

  • MDL number MFCD32068933
  • Cas Number : 2375274-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5928743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5928743
Availability details for
Quantity
  • Availability
  • Add to Cart