ENAH5802D9DE Enamine

3,6-dibromopyrazolo[1,5-a]pyrimidin-2-ol hydrochloride

  • MDL number MFCD32066068
  • Cas Number : 2361644-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432361
Availability details for
Quantity
  • Availability
  • Add to Cart