ENAH5802D9DF Enamine

3-(2,3-dimethylphenyl)-2-[2-(2-hydroxy-3-methoxyphenyl)ethenyl]-3,4-dihydroquinazolin-4-one

  • Cas Number : 936744-30-8
  • Molecular Weight : 398.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (Z)-3-(2,3-dimethylphenyl)-2-(2-hydroxy-3-methoxystyryl)quinazolin-4(3H)-one

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27049
Availability details for
Quantity
  • Availability
  • Add to Cart