ENAH5802D9E2 Enamine

2,2-difluorobicyclo[3.1.0]hexane-6-carbaldehyde

  • MDL number MFCD32069230
  • Cas Number : 2375260-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438917
Availability details for
Quantity
  • Availability
  • Add to Cart