ENAH5802D9E4 Enamine

5-bromo-2-cyclopropyl-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD32068728
  • Cas Number : 2375262-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427995
Availability details for
Quantity
  • Availability
  • Add to Cart