ENAH5802D9E7 Enamine

tert-butyl 4-(4-formyl-1,3-oxazol-5-yl)piperidine-1-carboxylate

  • MDL number MFCD32069284
  • Cas Number : 2375259-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7429765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429765
Availability details for
Quantity
  • Availability
  • Add to Cart