ENAH5802D9E8 Enamine

1-(but-3-yn-1-yl)-3-cyclopropylazetidin-3-ol

  • MDL number MFCD21329653
  • Cas Number : 1466790-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2479922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2479922
Availability details for
Quantity
  • Availability
  • Add to Cart