ENAH5802D9EB Enamine

1-(but-3-yn-1-yl)-3,3-dimethylpiperidine

  • Cas Number : 1340290-03-0
  • Molecular Weight : 165.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-butynyl)-3,3-dimethylpiperidine
Inchi Code 1S/C11H19N/c1-4-5-8-12-9-6-7-11(2,3)10-12/h1H,5-10H2,2-3H3
Inchi Key YNWGGNNCYZVPCH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3481664

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-3481664
pictograms GHS05,GHS07
hstatements H227,H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart