ENAH5802D9EC Enamine

3,3-dimethyl-1-(pent-4-yn-1-yl)pyrrolidine

  • MDL number MFCD26803897
  • Cas Number : 1566203-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5152533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5152533
Availability details for
Quantity
  • Availability
  • Add to Cart