ENAH5802D9ED Enamine

1-(but-3-yn-1-yl)-3,3-dimethylpiperazine

  • MDL number MFCD21112392
  • Cas Number : 1465215-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3039094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3039094
Availability details for
Quantity
  • Availability
  • Add to Cart