ENAH5802D9F2 Enamine

4-(but-3-yn-1-yl)-2,2-dimethylmorpholine

  • MDL number MFCD18851278
  • Cas Number : 1341194-07-7
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-butynyl)-2,2-dimethylmorpholine
Inchi Code 1S/C10H17NO/c1-4-5-6-11-7-8-12-10(2,3)9-11/h1H,5-9H2,2-3H3
Inchi Key YPTQOQUKOUFXHZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5067278

Safety Information

signalword Danger
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-5067278
pictograms GHS05,GHS07
hstatements H227,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart