ENAH5802D9F3 Enamine

1-(but-3-yn-1-yl)-3-methylpiperidin-3-ol

  • MDL number MFCD32068811
  • Cas Number : 1599226-56-8
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(but-3-yn-1-yl)-3-methylpiperidin-3-ol
Inchi Code 1S/C10H17NO/c1-3-4-7-11-8-5-6-10(2,12)9-11/h1,12H,4-9H2,2H3
Inchi Key YNSYCBGUKFTFBB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5171628

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-5171628
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart