ENAH5802D9F4 Enamine

1-(but-3-yn-1-yl)-4-methylpiperidin-4-ol

  • MDL number MFCD32066447
  • Cas Number : 1517358-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5179841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5179841
Availability details for
Quantity
  • Availability
  • Add to Cart