ENAH5802D9F6 Enamine

4-(but-3-yn-1-yl)-2-methylthiomorpholine

  • MDL number MFCD30072846
  • Cas Number : 1602597-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3472410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3472410
Availability details for
Quantity
  • Availability
  • Add to Cart