ENAH5802D9F7 Enamine

(2R)-4-(but-3-yn-1-yl)-2-methylthiomorpholine

  • MDL number MFCD32066519
  • Cas Number : 2361610-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6891965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6891965
Availability details for
Quantity
  • Availability
  • Add to Cart