ENAH5802D9FA Enamine

penta-2,4-diyn-1-amine hydrochloride

  • MDL number MFCD32065996
  • Cas Number : 2361635-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433906
Availability details for
Quantity
  • Availability
  • Add to Cart