ENAH5802D9FD Enamine

[3-(3-methoxyprop-1-yn-1-yl)phenyl]methanol

  • MDL number MFCD31924569
  • Cas Number : 2344680-20-0
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-(3-methoxyprop-1-yn-1-yl)phenyl)methanol
Inchi Code 1S/C11H12O2/c1-13-7-3-6-10-4-2-5-11(8-10)9-12/h2,4-5,8,12H,7,9H2,1H3
Inchi Key YXQZURSLRQTVJS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6526728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526728
Availability details for
Quantity
  • Availability
  • Add to Cart