ENAH5802D9FF Enamine

1-(but-3-yn-1-yl)-3-methylpiperidine-3-carbonitrile

  • MDL number MFCD32068970
  • Cas Number : 1882002-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3479999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3479999
Availability details for
Quantity
  • Availability
  • Add to Cart